MilliporeSigma
  • Superoxide Dismutase 3 Inhibits LL-37/KLK-5-Mediated Skin Inflammation through Modulation of EGFR and Associated Inflammatory Cascades.
Superoxide Dismutase 3 Inhibits LL-37/KLK-5-Mediated Skin Inflammation through Modulation of EGFR and Associated Inflammatory Cascades.
The Journal of investigative dermatology (2019-08-30)
Gaurav Agrahari, Shyam Kishor Sah, Cuong Thach Nguyen, Sung Sik Choi, Hae-Young Kim, Tae-Yoon Kim
ABSTRACT
The expressions of LL-37 and KLK-5 were found to be altered in various dermatoses, including atopic dermatitis, psoriasis, and rosacea. However, the downstream inflammatory effect of LL-37 and KLK-5 is not as well studied. In addition, there is little high-quality evidence for the treatment of LL-37- and KLK-5-mediated inflammation. In this study, we investigated the effect of superoxide dismutase 3 (SOD3) on LL-37- or KLK-5-induced skin inflammation in vitro and in vivo and its underlying anti-inflammatory mechanisms. Our data showed that SOD3 significantly reduced both LL-37- and KLK-5-induced expression of pro-inflammatory mediators and suppressed the activation of EGFR, protease-activated receptor 2, nucleotide-binding domain, leucine-rich-containing family, pyrin domain-containing-3, and p38/extracellular signal-regulated kinase signaling pathways in human keratinocytes. Moreover, SOD3 suppressed LL-37-induced expression of inflammatory mediators, reactive oxygen species production, and p38/extracellular signal-regulated kinase activation in mast cells. In addition, subcutaneous injection of KLK-5 in SOD3 knockout mice exhibited erythema with increased epidermal thickness, mast cell and neutrophil infiltration, expression of inflammatory mediators, and activation of EGFR, protease-activated receptor 2, nucleotide-binding domain, leucine-rich-containing family, pyrin domain-containing-3, and downstream mitogen-activated protein kinase pathways. However, treatment with SOD3 in SOD3 knockout mice rescued KLK-5-induced inflammatory cascades. Similarly, KLK-5-induced inflammation in wild-type mice was also ameliorated when treated with SOD3. Taken together, our data suggest that SOD3 is a potentially effective therapy for both LL-37-and KLK-5-induced skin inflammation.
Sigma-Aldrich
Sodium diethyldithiocarbamate trihydrate
Sigma-Aldrich
PD 153035, An extremely potent, cell-permeamble, reversible, ATP-competitive and specific inhibitor of the tyrosine kinase activity of the epidermal growth factor receptor (EGFR; IC₅₀ = 25 pM; Ki = 6 pM).