MilliporeSigma
  • C/EBPɑ is crucial determinant of epithelial maintenance by preventing epithelial-to-mesenchymal transition.
C/EBPɑ is crucial determinant of epithelial maintenance by preventing epithelial-to-mesenchymal transition.
Nature communications (2020-02-09)
Ana Rita Lourenço, M Guy Roukens, Danielle Seinstra, Cynthia L Frederiks, Cornelieke E Pals, Stephin J Vervoort, Andreia S Margarido, Jacco van Rheenen, Paul J Coffer
ABSTRACT
Extracellular signals such as TGF-β can induce epithelial-to-mesenchymal transition (EMT) in cancers of epithelial origin, promoting molecular and phenotypical changes resulting in pro-metastatic characteristics. We identified C/EBPα as one of the most TGF-β-mediated downregulated transcription factors in human mammary epithelial cells. C/EBPα expression prevents TGF-β-driven EMT by inhibiting expression of known EMT factors. Depletion of C/EBPα is sufficient to induce mesenchymal-like morphology and molecular features, while cells that had undergone TGF-β-induced EMT reverted to an epithelial-like state upon C/EBPα re-expression. In vivo, mice injected with C/EBPα-expressing breast tumor organoids display a dramatic reduction of metastatic lesions. Collectively, our results show that C/EBPα is required for maintaining epithelial homeostasis by repressing the expression of key mesenchymal markers, thereby preventing EMT-mediated tumorigenesis. These data suggest that C/EBPα is a master epithelial "gatekeeper" whose expression is required to prevent unwarranted mesenchymal transition, supporting an important role for EMT in mediating breast cancer metastasis.
Sigma-Aldrich
Insulin solution from bovine pancreas, 10 mg/mL insulin in 25  mM HEPES, pH 8.2, BioReagent, sterile-filtered, suitable for cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting human CEBPA
Sigma-Aldrich
MISSION® pLKO.1-puro Non-Mammalian shRNA Control Plasmid DNA, Targets no known mammalian genes