MilliporeSigma
  • Peel of Traditional Apple Varieties as a Great Source of Bioactive Compounds: Extraction by Micro-Matrix Solid-Phase Dispersion.
Peel of Traditional Apple Varieties as a Great Source of Bioactive Compounds: Extraction by Micro-Matrix Solid-Phase Dispersion.
Foods (Basel, Switzerland) (2020-01-17)
Ante Lončarić, Katarina Matanović, Perla Ferrer, Tihomir Kovač, Bojan Šarkanj, Martina Skendrović Babojelić, Marta Lores
ABSTRACT
Micro matrix solid phase dispersion (micro-MSPD) was optimized by response surface methodology for the extraction of polyphenols from the peel of twelve traditional and eight commercial apple varieties grown in Croatia. The optimized micro-MSPD procedure includes the use of 0.2 g of sample, 0.8 g of dispersant, a 57% solution of methanol in water as the solvent and 5 mL of extract volume. The total polyphenolic index (TPI) and antioxidant activity (AA) were measured by spectrophotometric assays. Eighteen polyphenolic compounds were identified in all investigated apples by HPLC-DAD and LC-(ESI)-MS. The peel of traditional apple varieties had higher contents of all investigated polyphenols. Calculated relative contribution of polyphenol groups indicated non-flavonoids (28.6%) and flavanols (46.2%) as the major contributors to the total polyphenolic content in traditional and commercial apple varieties, respectively. The most abundant polyphenol in traditional apple peel was chlorogenic acid, procyanidin B2 and epicatechin (1143 ± 755 µg/g dw, 954 ± 343 µg/g dw and 560 ± 362 µg/g dw, respectively). The peel of varieties 'Apistar', 'Bobovac' and 'Božićnica' could be highlighted as an important source of polyphenols.
Sigma-Aldrich
(±)-6-Hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid, 97%
Sigma-Aldrich
Gallic acid monohydrate, ≥99% (HPLC)
Sigma-Aldrich
Quercetin 3-β-D-glucoside, ≥90% (HPLC)