MilliporeSigma
  • Determination of peptide hormones of brain and intestine by CE with ESI-MS detection.
Determination of peptide hormones of brain and intestine by CE with ESI-MS detection.
Electrophoresis (2007-09-15)
Shifei Xia, Lan Zhang, Ping Tong, Minghua Lu, Wei Liu, Guonan Chen
ABSTRACT
A new method for the determination of the peptide hormones of brain and intestine based on CE coupling with a DAD and ESI-MS was established. Several electrophoretic and ESI-MS parameters were investigated in detail, such as electrolyte nature and concentration, organic solvent and sheath liquid compositions, nebulization gas pressure and the ESI capillary voltage. Optimized conditions were achieved with 25 mM formic acid-ammonium formate (pH 2.9) as the optimal electrolyte, 2 mM formic acid in 80% methanol in water as the sheath liquid, and 20 kV applied voltage. Under the optimized conditions, four protonated peptides were separated by CE and selectively detected by a quadrupole mass spectrometer with a sheath flow ESI interface. LODs for the four peptides (neurotensin hexapeptide, neurotensin, cholecystokinin tetrapeptide, and pentagastrin) were in the range of 0.10-0.60 micromol/L at an S/N of 3. The RSDs (n = 8) of the method were 0.70-1.5% for migration times and 1.6-6.1% for peak areas. This method is simple, rapid, and selective compared with RIA and ELISA techniques, and has been applied to the analysis of rat hypothalamus tissue.
Sigma-Aldrich
Neurotensin Fragment 8-13 acetate salt, ≥97% (HPLC)