MilliporeSigma
  • Activation of General Control Nonderepressible-2 Kinase Ameliorates Glucotoxicity in Human Peritoneal Mesothelial Cells, Preserves Their Integrity, and Prevents Mesothelial to Mesenchymal Transition.
Activation of General Control Nonderepressible-2 Kinase Ameliorates Glucotoxicity in Human Peritoneal Mesothelial Cells, Preserves Their Integrity, and Prevents Mesothelial to Mesenchymal Transition.
Biomolecules (2019-12-11)
Theodoros Eleftheriadis, Georgios Pissas, Georgia Antoniadi, Evdokia Nikolaou, Spyridon Golfinopoulos, Vassilios Liakopoulos, Ioannis Stefanidis
ABSTRACT
Along with infections, ultrafiltration failure due to the toxicity of glucose-containing peritoneal dialysis (PD) solutions is the Achilles' heel of PD method. Triggered by the protective effect of general control nonderepressible-2 (GCN-2) kinase activation against high-glucose conditions in other cell types, we evaluated whether the same occurs in human peritoneal mesothelial cells. We activated GCN-2 kinase with halofuginone or tryptophanol, and assessed the impact of this intervention on glucose transporter-1, glucose transporter-3, and sodium-glucose cotransporter-1, glucose influx, reactive oxygen species (ROS), and the events that result in glucotoxicity. These involve the inhibition of glyceraldehyde 3-phosphate dehydrogenase and the diversion of upstream glycolytic products to the aldose pathway (assessed by D-sorbitol), the lipid synthesis pathway (assessed by protein kinase C activity), the hexosamine pathway (determined by O-linked β-N-acetyl glucosamine-modified proteins), and the advanced glycation end products generation pathway (assessed by methylglyoxal). Then, we examined the production of the profibrotic transforming growth factor-β1 (TGF-β1), the pro-inflammatory interleukin-8 (IL-8). Cell apoptosis was assessed by cleaved caspase-3, and mesothelial to mesenchymal transition (MMT) was evaluated by α-smooth muscle actin protein. High-glucose conditions increased glucose transporters, glucose influx, ROS, all the high-glucose-induced harmful pathways, TGF-β1 and IL-8, cell apoptosis, and MMT. Halofuginone and tryptophanol inhibited all of the above high glucose-induced alterations, indicating that activation of GCN-2 kinase ameliorates glucotoxicity in human peritoneal mesothelial cells, preserves their integrity, and prevents MMT. Whether such a strategy could be applied in the clinic to avoid ultrafiltration failure in PD patients remains to be investigated.
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
L-Tryptophanol, 97%