MilliporeSigma
  • ProNGF correlates with Gleason score and is a potential driver of nerve infiltration in prostate cancer.
ProNGF correlates with Gleason score and is a potential driver of nerve infiltration in prostate cancer.
The American journal of pathology (2014-10-07)
Jay Pundavela, Yohann Demont, Phillip Jobling, Lisa F Lincz, Severine Roselli, Rick F Thorne, Danielle Bond, Ralph A Bradshaw, Marjorie M Walker, Hubert Hondermarck
ABSTRACT
Nerve infiltration is essential to prostate cancer progression, but the mechanism by which nerves are attracted to prostate tumors remains unknown. We report that the precursor of nerve growth factor (proNGF) is overexpressed in prostate cancer and involved in the ability of prostate cancer cells to induce axonogenesis. A series of 120 prostate cancer and benign prostate hyperplasia (BPH) samples were analyzed by IHC for proNGF. ProNGF was mainly localized in the cytoplasm of epithelial cells, with marked expression in cancer compared with BPH. Importantly, the proNGF level positively correlated with the Gleason score (n = 104, τB = 0.51). A higher level of proNGF was observed in tumors with a Gleason score of ≥8 compared with a Gleason score of 7 and 6 (P < 0.001). In vitro, proNGF was detected in LNCaP, DU145, and PC-3 prostate cancer cells and BPH-1 cells but not in RWPE-1 immortalized nontumorigenic prostate epithelial cells or primary normal prostate epithelial cells. Co-culture of PC12 neuronal-like cells or 50B11 neurons with PC-3 cells resulted in neurite outgrowth in neuronal cells that was inhibited by blocking antibodies against proNGF, indicating that prostate cancer cells can induce axonogenesis via secretion of proNGF. These data reveal that ProNGF is a biomarker associated with high-risk prostate cancers and a potential driver of infiltration by nerves.
Sigma-Aldrich
Anti-Nerve Growth Factor Antibody, pro, serum, Chemicon®