MilliporeSigma
  • MEASUREMENT OF URANIUM IN URINE, HAIR AND NAILS IN SUBJECTS OF NISKA BANJA TOWN, A HIGH NATURAL BACKGROUND RADIATION AREA OF SERBIA.
MEASUREMENT OF URANIUM IN URINE, HAIR AND NAILS IN SUBJECTS OF NISKA BANJA TOWN, A HIGH NATURAL BACKGROUND RADIATION AREA OF SERBIA.
Radiation protection dosimetry (2019-04-30)
Z S Žunic, L Benedik, R Kritsananuwat, N Veselinovic, S Tokonami, H Arae, Z Stojanovska, S K Sahoo
ABSTRACT
Urine, hair and nail samples were collected from a population of both sex aged from 40 to 87 years in Niška Banja (227 km south of Belgrade, Serbia) with a high level of natural radioactivity (HLNRA). To assess and monitor the public exposure, urine samples for each subject (24 h period) were collected and concentration of uranium (U) was analysed using alpha-particle spectrometry. Human hair and nails considered as bio-indicators of the public exposure to radionuclides over a long period of months or even years and concentration of U was determined using inductively coupled plasma-mass spectrometry. The mean concentrations of U in urine, hair and nail were 123 ng/L, 17 ng/g and 8 ng/g, respectively. There was a large variation and may be due to geological origin.
Sigma-Aldrich
Neodymium(III) fluoride, anhydrous, powder, 99.99% trace metals basis