MilliporeSigma
  • Imaging blood-brain barrier dysfunction as a biomarker for epileptogenesis.
Imaging blood-brain barrier dysfunction as a biomarker for epileptogenesis.
Brain : a journal of neurology (2017-04-27)
Guy Bar-Klein, Svetlana Lublinsky, Lyn Kamintsky, Iris Noyman, Ronel Veksler, Hotjensa Dalipaj, Vladimir V Senatorov, Evyatar Swissa, Dror Rosenbach, Netta Elazary, Dan Z Milikovsky, Nadav Milk, Michael Kassirer, Yossi Rosman, Yonatan Serlin, Arik Eisenkraft, Yoash Chassidim, Yisrael Parmet, Daniela Kaufer, Alon Friedman
ABSTRACT
A biomarker that will enable the identification of patients at high-risk for developing post-injury epilepsy is critically required. Microvascular pathology and related blood-brain barrier dysfunction and neuroinflammation were shown to be associated with epileptogenesis after injury. Here we used prospective, longitudinal magnetic resonance imaging to quantitatively follow blood-brain barrier pathology in rats following status epilepticus, late electrocorticography to identify epileptic animals and post-mortem immunohistochemistry to confirm blood-brain barrier dysfunction and neuroinflammation. Finally, to test the pharmacodynamic relevance of the proposed biomarker, two anti-epileptogenic interventions were used; isoflurane anaesthesia and losartan. Our results show that early blood-brain barrier pathology in the piriform network is a sensitive and specific predictor (area under the curve of 0.96, P < 0.0001) for epilepsy, while diffused pathology is associated with a lower risk. Early treatments with either isoflurane anaesthesia or losartan prevented early microvascular damage and late epilepsy. We suggest quantitative assessment of blood-brain barrier pathology as a clinically relevant predictive, diagnostic and pharmaco!dynamics biomarker for acquired epilepsy.
Sigma-Aldrich
Anti-Rat IgG (whole molecule)–FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Donkey Anti-Chicken IgY Antibody, Cy3 conjugate, Species Adsorbed, Chemicon®, from donkey