MilliporeSigma
  • Improved Stability and Photodynamic Activity of Water-Soluble 5,15-Diazaporphyrins Incorporated in β-(1,3-1,6)-d-Glucan with On-Off Switch.
Improved Stability and Photodynamic Activity of Water-Soluble 5,15-Diazaporphyrins Incorporated in β-(1,3-1,6)-d-Glucan with On-Off Switch.
Chemistry, an Asian journal (2019-12-13)
Shodai Hino, Shuhei Satake, Hideyuki Shinmori, Shigeki Kawabata, Kazuya Koumoto, Toshio Suzuki, Takeshi Nagasaki, Kouta Sugikawa, Riku Kawasaki, Atsushi Ikeda
ABSTRACT
5,15-Diazaporphyrins, which have a large absorption at wavelengths over 600 nm, were dissolved in water by complex formation with β-(1,3-1,6)-d-glucans. Aqueous solutions of these complexes were relatively stable compared with their trimethyl-β-cyclodextrin-complexed analogues. β-Glucan-complexed diazaporphyrins showed quenched fluorescence and had low singlet-oxygen-generation abilities owing to random self-aggregation. However, external stimuli, such as the presence of liposomes or intracellular uptake, restored the fluorescence and singlet-oxygen-generation abilities of β-glucan-complexed diazaporphyrins. Consequently, β-glucan-complexed diazaporphyrins showed very high photodynamic activities toward HeLa cells.
Sigma-Aldrich
9,10-Anthracenediyl-bis(methylene)dimalonic acid, BioReagent, suitable for fluorescence, ≥90% (HPCE)