MilliporeSigma
  • Differential effects of phosphonic analogues of GABA on GABA(B) autoreceptors in rat neocortical slices.
Differential effects of phosphonic analogues of GABA on GABA(B) autoreceptors in rat neocortical slices.
Naunyn-Schmiedeberg's archives of pharmacology (1998-05-30)
J Ong, V Marino, D A Parker, D I Kerr
ABSTRACT
The effects of five phosphonic derivatives of GABA on the release of [3H]-GABA from rat neocortical slices, preloaded with [3H]-GABA, were investigated. Phaclofen and 4-aminobutylphosphonic acid (4-ABPA) increased the overflow of [3H] evoked by electrical stimulation (2 Hz) in a concentration-dependent manner, with similar potencies (phaclofen EC50=0.3 mmol/l, 4-ABPA EC50=0.4 mmol/l). At 3 mmol/l, phaclofen increased the release of [3H]-GABA by 82.6+/-8.6%, and 4-ABPA increased the release by 81.3+/-9.0%. 2-Amino-ethylphosphonic acid (2-AEPA) increased the overflow of [3H] by 46.8+/-10.9% at the highest concentration tested (3 mmol/l). In contrast, the lower phosphonic homologue 3-aminopropylphosphonic acid (3-APPA), and 2-amino-2-(p-chlorophenyl)-ethylphosphonic acid (2-CPEPA), a baclofen analogue, did not modify the stimulated overflow. These results suggest that phaclofen, 4-ABPA and 2-AEPA are antagonists at GABA(B) autoreceptors, the latter being the weakest antagonist, whilst neither 3-APPA nor 2-CPEPA are active at these receptors. Since phaclofen, 4-ABPA and 2-CPEPA are antagonists and 3-APPA a partial agonist/antagonist on GABA(B) heteroreceptors, the lack of effect of 3-APPA and 2-CPEPA on [3H]-GABA release in this study suggests that GABA(B) autoreceptors may be pharmacologically distinct from the heteroreceptors.
Sigma-Aldrich
4-Aminobutylphosphonic acid, ≥99%
Sigma-Aldrich
3-Aminopropylphosphonic acid, 98%