MilliporeSigma
  • Exploiting Synergistic Catalysis for an Ambient Temperature Photocycloaddition to Pyrazoles.
Exploiting Synergistic Catalysis for an Ambient Temperature Photocycloaddition to Pyrazoles.
Chemistry (Weinheim an der Bergstrasse, Germany) (2019-10-28)
Christopher P Lakeland, David W Watson, Joseph P A Harrity
ABSTRACT
Sydnone-based cycloaddition reactions are a versatile platform for pyrazole synthesis, however they operate under harsh conditions (high temperature and long reaction times). Herein we report a strategy that addresses this limitation utilizing the synergistic combination of organocatalysis and visible-light photocatalysis. This new approach proceeds under ambient conditions and with excellent levels of regiocontrol. Mechanistic studies suggest that photoactivation of sydnones, rather than enamines, is key to the successful implementation of this process.