MilliporeSigma
  • TCF19 Promotes Cell Proliferation through Binding to the Histone H3K4me3 Mark.
TCF19 Promotes Cell Proliferation through Binding to the Histone H3K4me3 Mark.
Biochemistry (2019-11-21)
Payel Mondal, Sabyasachi Sen, Brianna J Klein, Niharika Tiwary, Shrikanth S Gadad, Tatiana G Kutateladze, Siddhartha Roy, Chandrima Das
ABSTRACT
Transcription factor 19 (TCF19) plays critical roles in type 1 diabetes and the maintenance of pancreatic β cells. Recent studies have also implicated TCF19 in cell proliferation of hepatic carcinoma and non-small cell lung carcinoma; however, the mechanism underlying this regulation remains elusive. At the molecular level, TCF19 contains two modules, the plant homeodomain (PHD) finger and the forkhead-associated (FHA) domain, of unclear function. Here, we show that TCF19 mediates hepatocellular carcinoma HepG2 cell proliferation through its PHD finger that recognizes trimethylated lysine 4 of histone 3 (H3K4me3). W316 of the PHD finger of TCF19 is one of the critical residues eliciting this function. Whole genome microarray analysis and orthogonal cell-based assays identified a large subset of genes involved in cell survival and proliferation that depend on TCF19. Our data suggest that TCF19 acts as a pro-oncogene in hepatocellular carcinoma cells and that its functional PHD finger is critical in cell proliferation.
Sigma-Aldrich
MISSION® esiRNA, targeting human TCF19