MilliporeSigma
  • Inhibition of kras-derived exosomes downregulates immunosuppressive BACH2/GATA-3 expression via RIP-3 dependent necroptosis and miR-146/miR-210 modulation.
Inhibition of kras-derived exosomes downregulates immunosuppressive BACH2/GATA-3 expression via RIP-3 dependent necroptosis and miR-146/miR-210 modulation.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2020-01-10)
Savvas Petanidis, Kalliopi Domvri, Konstantinos Porpodis, Doxakis Anestakis, Lutz Freitag, Wolfgang Hohenforst-Schmidt, Drosos Tsavlis, Konstantinos Zarogoulidis
ABSTRACT
Immunosuppressive chemoresistance is a major challenge in lung cancer treatment. Exosomes present in the tumor microenviroment are implicated in chemoresistant-related immune suppression, and metastasis but the exact pathogenic role of lung-derived exosomes is still uncertain. Recent reports reveal that lung cancer pathogenesis is strictly associated with a exosomal tumor supportive status and a dysfunctional immune system. In this study, we investigate the role of Kras-derived exosomes in chemoresistant immunosuppression in which neoplastic cells create a metabolic-sustained microenvironment. Findings reveal that Kras-derived exosomes induce regulation of SMARCE1/NCOR1 chromatin remodeling genes promoting pre-metastatic niche formation in naive mice and consequently increase lung metastatic burden. Furthermore, exosomal Kras inhibition downregulated transcription factor BACH2/GATA-3 expression in lung tumor tissues by shifting pyruvate/PKM2 dependent metabolism, contributing to a tumor-restraining status. Further co-treatment with carboplatin triggered RIP3/TNFa dependent necroptosis in ex vivo cells accompanied by differential expression of immunosuppressive miR-146/miR-210 regulators in metastatic lung cancer patients. Overall, these findings demonstrate the multifaceted roles of Kras-derived exosomes in sustaining lung immunosuppressive metastasis and provide new opportunities for effective metastasis inhibition, especially in chemoresistant tumors.
Sigma-Aldrich
MISSION® esiRNA, targeting human SMARCE1
Sigma-Aldrich
MISSION® esiRNA, targeting human KRAS