MilliporeSigma
  • Silencing of TXNIP Alleviated Oxidative Stress Injury by Regulating MAPK-Nrf2 Axis in Ischemic Stroke.
Silencing of TXNIP Alleviated Oxidative Stress Injury by Regulating MAPK-Nrf2 Axis in Ischemic Stroke.
Neurochemical research (2019-12-21)
Yu Tian, Yue Su, Qile Ye, Lei Chen, Fei Yuan, Zhenyu Wang
ABSTRACT
Ischemic stroke is a life-threatening cerebrovascular thrombotic disease, oxidative stress is considered to be a critical factor to stroke pathophysiology. This study aimed to investigate the underlying molecular mechanism and propose the potential therapeutic strategy for ischemic stroke. Bioinformatics analysis based on a public microarray profile (GSE 61616) of ischemic stroke rats was performed as a pilot research. Oxidative stress was enriched as a significantly gene ontology item, and thioredoxin-interacting protein (TXNIP) and MAPK signaling were identified as the hub gene and pathway, respectively. The experiments in middle cerebral artery occlusion rats demonstrated that ischemia induced the activation of oxidative stress. The expressions of TXNIP, p-p38, p-JNK, p-ERK were significantly increased while Nrf2 and HO-1 expressions were decreased after stroke. Rescue assays were conducted in primary cultured neurons to explore the accurate interrelations among these factors. The results indicated that MAPK specific inhibitor and siRNA-TXNIP significantly alleviated the oxidative stress injury induced by oxygen-glucose deprivation. In addition, knocking down of TXNIP inhibited the activation of MAPK pathway and promoted Nrf2 pathway. Taken together, these findings indicated that TXNIP aggravated the oxidative stress injury by regulating MAPK-Nrf2 axis in ischemic stroke. Silencing of TXNIP seems a promising therapeutic strategy to alleviate ischemic stroke.
Sigma-Aldrich
MISSION® esiRNA, targeting human TXNIP