MilliporeSigma
  • PICK1 attenuates high glucose-induced pancreatic β-cell death through the PI3K/Akt pathway and is negatively regulated by miR-139-5p.
PICK1 attenuates high glucose-induced pancreatic β-cell death through the PI3K/Akt pathway and is negatively regulated by miR-139-5p.
Biochemical and biophysical research communications (2019-11-19)
Haiyan Qiu, Lizhen Ma, Fabo Feng
ABSTRACT
Diabetes mellitus is a metabolic disease characterized by an increase in blood glucose levels due to lack of insulin secretion. Previous studies have confirmed that PICK1 is critical for both β-cell function and glucose homeostasis. The aim of this study was to investigate the role of PICK1 in response to high glucose-induced β-cell dysfunction and the molecular mechanism of regulation of PICK1. We found that overexpression of PICK1 in db/db diabetic mice significantly improved glucose tolerance and increased insulin release. High glucose treatment of Min6 cells inhibited PICK1 expression, and overexpression of PICK1 protected against high glucose-induced pancreatic cell dysfunction. Activation of the PI3K/Akt pathway by PICK1 in Min6 cells resulted in increased GLUT2 expression and this increase was abolished by treatment with a PI3K-specific inhibitor. Further, we showed that expression of PICK1 is negatively regulated by miR-139-5p through directly targeting its 3'UTR. These data suggested that PICK1 may participate in the functional protection of pancreatic β-cells through PI3K/Akt signaling, promote insulin secretion, and delay the progression of diabetes, and is negatively regulated by miR-139-5p, further clarifying the regulation of pancreatic β-cell function.
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder