MilliporeSigma
Derlin-1 facilitates the retro-translocation of cholera toxin.
Molecular biology of the cell (2007-12-21)
Kaleena M Bernardi, Michele L Forster, Wayne I Lencer, Billy Tsai
ABSTRACT
Cholera toxin (CT) intoxicates cells by using its receptor-binding B subunit (CTB) to traffic from the plasma membrane to the endoplasmic reticulum (ER). In this compartment, the catalytic A1 subunit (CTA1) is unfolded by protein disulfide isomerase (PDI) and retro-translocated to the cytosol where it triggers a signaling cascade, leading to secretory diarrhea. How CT is targeted to the site of retro-translocation in the ER membrane to initiate translocation is unclear. Using a semipermeabilized-cell retro-translocation assay, we demonstrate that a dominant-negative Derlin-1-YFP fusion protein attenuates the ER-to-cytosol transport of CTA1. Derlin-1 interacts with CTB and the ER chaperone PDI as assessed by coimmunoprecipitation experiments. An in vitro membrane-binding assay showed that CTB stimulated the unfolded CTA1 chain to bind to the ER membrane. Moreover, intoxication of intact cells with CTB stabilized the degradation of a Derlin-1-dependent substrate, suggesting that CT uses the Derlin-1 pathway. These findings indicate that Derlin-1 facilitates the retro-translocation of CT. CTB may play a role in this process by targeting the holotoxin to Derlin-1, enabling the Derlin-1-bound PDI to unfold the A1 subunit and prepare it for transport.