MilliporeSigma
  • Antineuropathic pain actions of Wu-tou decoction resulted from the increase of neurotrophic factor and decrease of CCR5 expression in primary rat glial cells.
Antineuropathic pain actions of Wu-tou decoction resulted from the increase of neurotrophic factor and decrease of CCR5 expression in primary rat glial cells.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2020-01-17)
Qiuyan Guo, Koji Mizuno, Katsuki Okuyama, Na Lin, Yanqiong Zhang, Hideki Hayashi, Norio Takagi, Takashi Sato
ABSTRACT
Wu-tou decoction (WTD), a classic Traditional Chinese medicine formula, has been extensively used in the treatment of neuropathic pain (NP) such as chronic inflammatory pain, trigeminal neuralgia, and cancer-induced pain. Our previous studies have shown that the severity of mechanical allodynia and thermo hypersensitivity in NP rats are reduced by WTD, of which analgesic candidates are paeoniflorin (Pae) and liquiritin (Liq). The aim of this study was to clarify the molecular mechanisms of WTD, Pae and Liq against NP based on the primary rat glial cells in vitro. The gene expression levels of neurotrophic factors such as nerve growth factor (NGF), brain-derived neurotrophic factor (BDNF), glial cell line-derived neurotrophic factor (GDNF), and Artemin and C-C chemokine receptor type 5 (CCR5) were augmented by inflammatory cytokines, while chemokines increased only CCR5 gene expression. The constitutive and cytokine-augmented neurotrophic factor gene expression was enhanced by WTD, Pae, and Liq through PI3K- and PKA-dependent pathways in rat glial cells, leading to the increase of NGF and BDNF production. Furthermore, the CCR5 gene expression under basal and chemokine-treated conditions was suppressed by these reagents, in which signal pathway(s) was independent on the activation of PI3K and PKA. Moreover, there was no cytotoxicity in the WTD, Pae, and Liq treatments in glial cells. Thus, these results provide a novel evidence that WTD may exert the anti-NP actions by predominantly increasing the production of neurotrophic factors through PI3K- and PKA-signaling pathways in rat glial cells. Furthermore, Pae and Liq may play as analgesic candidates in WTD-mediated NP management.
Sigma-Aldrich
Liquiritin, from Glycyrrhizae root (licorice root), ≥98% (HPLC)