MilliporeSigma
  • Biocompatible Organic Coatings Based on Bisphosphonic Acid RGD-Derivatives for PEO-Modified Titanium Implants.
Biocompatible Organic Coatings Based on Bisphosphonic Acid RGD-Derivatives for PEO-Modified Titanium Implants.
Molecules (Basel, Switzerland) (2020-01-16)
Lyudmila V Parfenova, Elena S Lukina, Zulfia R Galimshina, Guzel U Gil'fanova, Veta R Mukaeva, Ruzil G Farrakhov, Ksenia V Danilko, Grigory S Dyakonov, Evgeny V Parfenov
ABSTRACT
Currently, significant attention is attracted to the problem of the development of the specific architecture and composition of the surface layer in order to control the biocompatibility of implants made of titanium and its alloys. The titanium surface properties can be tuned both by creating an inorganic sublayer with the desired morphology and by organic top coating contributing to bioactivity. In this work, we developed a composite biologically active coatings based on hybrid molecules obtained by chemical cross-linking of amino acid bisphosphonates with a linear tripeptide RGD, in combination with inorganic porous sublayer created on titanium by plasma electrolytic oxidation (PEO). After the addition of organic molecules, the PEO coated surface gets nobler, but corrosion currents increase. In vitro studies on proliferation and viability of fibroblasts, mesenchymal stem cells and osteoblast-like cells showed the significant dependence of the molecule bioactivity on the structure of bisphosphonate anchor and the linker. Several RGD-modified bisphosphonates of β-alanine, γ-aminobutyric and ε-aminocaproic acids with BMPS or SMCC linkers can be recommended as promising candidates for further in vivo research.
Sigma-Aldrich
N-Hydroxysuccinimide, 98%