MilliporeSigma
  • Steroid hormone 20-hydroxyecdysone induces the transcription and complex assembly of V-ATPases to facilitate autophagy in Bombyx mori.
Steroid hormone 20-hydroxyecdysone induces the transcription and complex assembly of V-ATPases to facilitate autophagy in Bombyx mori.
Insect biochemistry and molecular biology (2019-10-28)
Yichen Dai, Kang Li, Wenmei Wu, Kunzhong Wu, Huiyu Yi, Wangyu Li, Yang Xiao, Yangjin Zhong, Yang Cao, Ling Tian
ABSTRACT
Vacuolar-type H + -adenosine triphosphatases (V-ATPases) are indispensable for lysosome acidification and participate in autophagic processes. The steroid hormone 20-hydroxyecdysone (20E) predominantly induces autophagy and regulates insect larval molting and metamorphosis; however, the specific mechanism of lysosome acidification regulation by 20E remains unclear. Here, we showed that the developmental profiles of Bombyx V-ATPases were in accordance with autophagy occurrence and lysosome acidification in the fat body during larval-pupal metamorphosis. BmV-ATPase-A and BmV-ATPase-B were required for lysosome acidification and autophagic flux. Both 20E treatment and starvation were able to induce lysosome acidification. Furthermore, BmV-ATPase transcription was induced by 20E treatment and reduced by RNAi targeting the 20E receptor BmUsp. On the one hand, 20E upregulated the transcription of BmV-ATPases through inducing Bombyx transcription factor EB (TFEB) and its nuclear translocation; on the other hand, 20E inhibited mTOR signaling to induce the transcription and assembly of BmV-ATPase subunits. Overall, 20E induces lysosome acidification by upregulating the transcription and assembly of V-ATPase subunits via activating BmTFEB and cooperating with nutrient signaling. These findings improve our understanding of the regulatory mechanisms underlying lysosome acidification and autophagic flux in Bombyx mori.
Sigma-Aldrich
20-Hydroxyecdysone, ≥93% (HPLC), powder
Sigma-Aldrich
Grace′s Insect Medium, With L-glutamine, without sodium bicarbonate, powder, suitable for insect cell culture