MilliporeSigma
  • MicroRNA-200c-3p inhibits proliferation and migration of renal artery endothelial cells by directly targeting ZEB2.
MicroRNA-200c-3p inhibits proliferation and migration of renal artery endothelial cells by directly targeting ZEB2.
Experimental cell research (2019-12-28)
Yao Liu, Yuehua Jiang, Wei Li, Cong Han, Le Zhou, Hongzhen Hu
ABSTRACT
Continuous activation of angiotensin II (Ang II) induces renal vascular endothelial dysfunction, inflammation, and oxidative stress, all of which may contribute to renal damage. It is well established that microRNAs (miRNAs) play crucial regulatory roles in the pathogenesis of hypertensive renal damage. However, the detailed mechanisms and regulatory roles of miRNAs as therapeutic targets underlying Ang II-induced renal artery endothelial cell dysfunction in hypertensive renal damage have yet to be fully elucidated. The present study aimed to explore the expression status and putative role of miRNA-200c-3p in mediating the progression of hypertensive renal damage. We carried out real-time quantitative PCR (RT-qPCR) to detect the expression of miRNA-200c-3p in rat renal artery endothelial cells (RRAECs) induced by Ang II. MTT and transwell assays were utilized to evaluate the effects of miRNA-200c-3p on cell proliferation and migration, respectively. The present results revealed that the expression of miRNA-200c-3p was significantly upregulated in RRAECs exposed to Ang II compared with that of normal cells. miRNA-200c-3p overexpression markedly inhibited cell proliferation and migration of Ang II-induced RRAECs. Furthermore, bioinformatics predictions and dual-luciferase reporter assays indicated that zinc finger E-box-binding homeobox 2 (ZEB2) was a direct target gene of miRNA-200c-3p and that ZEB2 expression was inversely correlated with the levels of miRNA-200c-3p in RRAECs after exposure to Ang II. The effects of ZEB2 silencing were similar to the inhibitory effects observed following miRNA-200c-3p overexpression, and recovered ZEB2 expression reversed the anti-proliferative and anti-migratory influence of miRNA-200c-3p upregulation in RRAECs induced by Ang II. The present study indicated that miRNA-200c-3p might suppress the proliferation and migration of Ang II-induced RRAECs by targeting ZEB2. The miRNA-200c-3p/ZEB2 axis will provide valuable insights into the clinical management of hypertension-related kidney disease.
Sigma-Aldrich
MISSION® esiRNA, targeting human CHN1
Sigma-Aldrich
MISSION® esiRNA, targeting human ZEB2