MilliporeSigma
  • Repetitive DNA landscape in essential A and supernumerary B chromosomes of Festuca pratensis Huds.
Repetitive DNA landscape in essential A and supernumerary B chromosomes of Festuca pratensis Huds.
Scientific reports (2019-12-29)
Rahman Ebrahimzadegan, Andreas Houben, Ghader Mirzaghaderi
ABSTRACT
Here, we characterized the basic properties of repetitive sequences in essential A and supernumerary B chromosomes of Festuca pratensis Huds. This was performed by comparative analysis of low-pass Illumina sequence reads of B chromosome lacking (-B) and B chromosome containing (+B) individuals of F. pratensis. 61% of the nuclear genome is composed of repetitive sequences. 43.1% of the genome are transposons of which DNA transposons and retrotransposons made up 2.3% and 40.8%, respectively. LTR retrotransposons are the most abundant mobile elements and contribute to 40.7% of the genome and divided into Ty3-gypsy and Ty1-copia super families with 32.97% and 7.78% of the genome, respectively. Eighteen different satellite repeats were identified making up 3.9% of the genome. Five satellite repeats were used as cytological markers for chromosome identification and genome analysis in the genus Festuca. Four satellite repeats were identified on B chromosomes among which Fp-Sat48 and Fp-Sat253 were specific to the B chromosome of F. pratensis.
Product Description
Millipore
CELLULYSIN Cellulase, Trichoderma viride
Sigma-Aldrich
Cytohelicase from Helix pomatia, lyophilized powder
Sigma-Aldrich
Pectolyase from Aspergillus japonicus, lyophilized powder, ≥0.3 units/mg solid