MilliporeSigma
  • Novel Biochemical Markers of Neurovascular Complications in Type 1 Diabetes Patients.
Novel Biochemical Markers of Neurovascular Complications in Type 1 Diabetes Patients.
Journal of clinical medicine (2020-01-16)
Bogusz Falkowski, Anita Rogowicz-Frontczak, Ewelina Szczepanek-Parulska, Aleksandra Krygier, Elzbieta Wrotkowska, Aleksandra Uruska, Aleksandra Araszkiewicz, Marek Ruchala, Dorota Zozulinska-Ziolkiewicz
ABSTRACT
Type 1 diabetes mellitus (T1DM) is associated with chronic complications, which are the result of neurovascular changes. There is still a lack of universal biochemical markers of microvascular damage. The present study aimed to investigate whether selected inflammatory proteins are related to the prevalence of microvascular complications in adult T1DM patients. The following markers were determined in a group of 100 T1DM participants: epidermal growth factor (EGF), metalloproteinase 2 (MMP-2), growth/differentiation factor 15 (GDF-15), and interleukin 29 (IL-29). Screening for microvascular complications, such as autonomic and peripheral neuropathy, diabetic kidney disease, and retinopathy, was conducted. The group was divided according to the occurrence of microvascular complications. At least one complication was required for the patient to be included in the microangiopathy group. The median EGF concentration in the microangiopathy group was higher than in the group without microangiopathy (p = 0.03). Increasing EGF concentration was a statistically significant predictor of the presence of microangiopathy in multivariate logistic regression analysis (p < 0.0001). Additionally, a higher GDF-15 level was associated with diabetic kidney disease, peripheral neuropathy, and proliferative retinopathy vs. nonproliferative retinopathy. GDF-15 concentration correlated negatively with estimated glomerular filtration rate (eGFR) (r = -0.28; p = 0.02). To conclude, higher EGF concentration is an independent predictor of the presence of microvascular complications in T1DM patients. Besides the relation between GDF-15 and diabetic kidney disease, it may be also associated with peripheral neuropathy and retinopathy.
Sigma-Aldrich
Human IFNL1 / Interferon lambda-1 ELISA Kit, for serum, plasma, cell culture supernatant and urine