MilliporeSigma
  • TRAF3 enhances TRIF-mediated signaling via NF-κB and IRF3 activation in large yellow croaker Larimichthys crocea.
TRAF3 enhances TRIF-mediated signaling via NF-κB and IRF3 activation in large yellow croaker Larimichthys crocea.
Fish & shellfish immunology (2019-12-17)
Peng Fei Zou, Juan Juan Shen, Ying Li, Zi Ping Zhang, Yi Lei Wang
ABSTRACT
As a member of tumor necrosis factor receptor (TNFR)-associated factor (TRAF) family, TRAF3 is an important regulator of NF-κB and type I interferon (IFN) activation, especially in Toll-like receptors (TLRs)- and retinoic acid inducible gene I (RIG-I)-like receptors (RLRs)-mediated signaling pathway. In the present study, a TRAF3 homologue named Lc-TRAF3 was characterized in large yellow croaker (Larimichthys crocea). The open reading frame (ORF) of Lc-TRAF3 contains 1788 bp encoding a protein of 595 amino acids (aa). Sequence analysis indicated that Lc-TRAF3 is conserved in vertebrates, constituted with a N-terminal RING finger, two TRAF-type zinc fingers, and a C-terminal TRAF-MATH domain. The genome organization of Lc-TRAF3 is conserved in fish, with 13 exons and 12 introns, but different from that in birds or mammals, which contains 10 exons and 9 introns. Lc-TRAF3 was identified as cytosolic protein base on fluorescence microscopy analysis. Expression analysis revealed that Lc-TRAF3 was broadly distributed in examined organs/tissues, with the highest expression level in gill and weakest in brain, and could be up-regulated under poly I:C, LPS, PGN, and Pseudomonas plecoglossicida stimulation in vivo. Interestingly, overexpression Lc-TRAF3 could induce the activation of NF-κB, and Lc-TRAF3 co-transfected with Lc-TRIF induced a significantly higher level of NF-κB and IRF3 promoter activity, implying that Lc-TRAF3 may function as an enhancer in Lc-TRIF-mediated signaling pathway.
Sigma-Aldrich
Peptidoglycan from Bacillus subtilis
Sigma-Aldrich
Polyinosinic–polycytidylic acid potassium salt, with buffer salts, TLR ligand tested
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, purified by ion-exchange chromatography, TLR ligand tested