MilliporeSigma
  • Determination of color developers replacing bisphenol A in thermal paper receipts using diode array and Corona charged aerosol detection-A German market analysis 2018/2019.
Determination of color developers replacing bisphenol A in thermal paper receipts using diode array and Corona charged aerosol detection-A German market analysis 2018/2019.
Journal of chromatography. A (2019-08-15)
Martin Eckardt, Marie Kubicova, Duyen Tong, Thomas J Simat
ABSTRACT
Thermal papers (e.g. point of sale receipts, adhesive labels, tickets) significantly contribute to contamination of paper material cycles and the environment with substances of (eco-) toxicological concern. In particular, they contain color developers like endocrine disrupting bisphenols in typical concentrations of about 1-2 percent per weight (wt%). Bisphenol A (BPA) was used as the common color developer over the last decades, but it will be restricted for thermal paper application in the European Union to a limit of 0.02 wt% from 2020 onwards. Consequently, a variety of BPA substituents such as bisphenol S (BPS) and its derivatives gain importance in thermal paper application. In this study, a rapid, reliable and cost-effective method for identification and quantification of BPA, alternative color developers and related substances like sensitizers is presented based on HPLC separation coupled with diode array detection (DAD) and Corona charged aerosol detection (CAD). Quantification was performed with regard to the intended use of the substances in thermal papers. Besides traditional UV external calibration using reference standards, alternative quantification approaches, in particular UV chromophore concentration for BPS derivatives and CAD universal response technique for low-volatile color developers, were applied and compared in order to allow quantification without reference substances. A market analysis for intended used color developers and sensitizers was performed on thermal paper samples (n = 211) collected in Germany during 2018 and 2019. Pergafast 201 (in 41.7% of the samples) was the most common color developer with concentrations above 0.02 wt%, followed by BPA (36.0%), BPS (13.3%) and other BPS derivatives known as D8, D-90, BPS-MAE and TGSA, that are mainly present in adhesive labels. Sensitizers were determined in over 90% of the samples.
Sigma-Aldrich
1,2-Diphenoxyethane, 99%
Sigma-Aldrich
4,4′-Sulfonyldiphenol, 98%