MilliporeSigma
  • Oxidative inactivation of amyloid beta-degrading proteases by cholesterol-enhanced mitochondrial stress.
Oxidative inactivation of amyloid beta-degrading proteases by cholesterol-enhanced mitochondrial stress.
Redox biology (2019-08-04)
Cristina de Dios, Isabel Bartolessis, Vicente Roca-Agujetas, Elisabet Barbero-Camps, Montserrat Mari, Albert Morales, Anna Colell
ABSTRACT
Familial early-onset forms of Alzheimer's disease (AD) are linked to overproduction of amyloid beta (Aβ) peptides, while decreased clearance of Aβ is the driving force leading to its toxic accumulation in late-onset (sporadic) AD. Oxidative modifications and defective function have been reported in Aβ-degrading proteases such as neprilysin (NEP) and insulin-degrading enzyme (IDE). However, the exact mechanisms that regulate the proteolytic clearance of Aβ and its deficits are largely unknown. We have previously showed that cellular cholesterol loading, by depleting the mitochondrial GSH (mGSH) content, stimulates Αβ-induced mitochondrial oxidative stress and promotes AD-like pathology in APP-PSEN1-SREBF2 mice. Here, using the same AD mouse model we examined whether cholesterol-enhanced mitochondrial oxidative stress affects NEP and IDE function. We found that brain extracts from APP-PSEN1-SREBF2 mice displayed increased presence of oxidatively modified forms of NEP and IDE, associated with impaired enzymatic activities. Both alterations were substantially recovered after an in vivo treatment with the cholesterol-lowering agent 2-hydroxypropyl-β-cyclodextrin. The recovery of the proteolytic activity after treatment was accompanied with a significant reduction of Aβ levels. Supporting these results, cholesterol-enriched SH-SY5Y cells were more sensitive to Aβ-induced impairment of IDE and NEP function in vitro. The rise of cellular cholesterol also stimulated the extracellular release of IDE by an unconventional autophagy-coordinated mechanism. Recovery of depleted pool of mGSH in these cells not only prevented the detrimental effect of Aβ on intracellular AβDPs activities but also had an impact on extracellular IDE levels and function, stimulating the extracellular Aβ degrading activity. Therefore, changes in brain cholesterol levels by modifying the mGSH content would play a key role in IDE and NEP-mediated proteolytic elimination of Aβ peptides and AD progression.
Sigma-Aldrich
Anti-Neprilysin Antibody (Neutral Endopeptidase, Nep), serum, Chemicon®
Sigma-Aldrich
Anti-IDE, N-Terminal (97-273) Rabbit pAb, liquid, Calbiochem®