MilliporeSigma
  • Two Four-Coordinate and Seven-Coordinate CoII Complexes Based on the Bidentate Ligand 1, 8-Naphthyridine Showing Slow Magnetic Relaxation Behavior.
Two Four-Coordinate and Seven-Coordinate CoII Complexes Based on the Bidentate Ligand 1, 8-Naphthyridine Showing Slow Magnetic Relaxation Behavior.
Chemistry, an Asian journal (2019-12-04)
Xing-Cai Huang, Rui Xu, Yong-Zhi Chen, Yi-Quan Zhang, Dong Shao
ABSTRACT
For a long time, the cobalt(II) complex ([Co(napy)4 ](ClO4 )2 ) (napy=1, 8-naphthyridine) has been considered as an eight-coordinate complex without any structural proof. After careful considerations, two complexes [Co(napy)2 Cl2 ] (1) and [Co(napy)4 ](ClO4 )2 (2) based on the bidentate ligand napy were synthesized and structurally characterized. X-ray single-crystal structural determination showed that the cobalt(II) center in [Co(napy)2 Cl2 ] (1) is four-coordinate with a tetrahedral geometry (Td ), while [Co(napy)4 ](ClO4 )2 (2) is seven-coordinate rather than eight-coordinate with a capped trigonal prism geometry (C2v ). Direct-current (dc) magnetic data revealed that complexes 1 and 2 possess positive zero-field splitting (ZFS) parameters of 11.08 and 25.30 cm-1 , respectively, with easy-plane magnetic anisotropy. Alternating current(ac) susceptibility measurements revealed that both complexes showed slow magnetic relaxation behaviour. Theoretical calculations demonstrated that the presence of easy-plane magnetic anisotropy (D>0) for complexes 1 and 2 is in agreement with the experimental data. Furthermore, these results pave the way to obtain four-coordinate and seven-coordinate cobalt(II) single-ion magnets (SIMs) by using a bidentate ligand.
Sigma-Aldrich
1,8-Naphthyridine