MilliporeSigma
  • The effects of crocin, mesalazine and their combination in the acetic acid-induced colitis in rats.
The effects of crocin, mesalazine and their combination in the acetic acid-induced colitis in rats.
Veterinary research forum : an international quarterly journal (2019-11-19)
Amir Faramarzpour, Ali Asghar Tehrani, Esmaeal Tamaddonfard, Mehdi Imani
ABSTRACT
Crocin, as a carotenoid compound of saffron, exerts a potent antioxidant property. Mesalazine is frequently used in the treatment of ulcerative colitis. This study investigated the effects of separated and combination treatments with crocin and mesalazine in a rat model of ulcerative colitis. Ulcerative colitis was induced by intra-colonic administration of acetic acid (4.00%, 1.00 mL) at 8 cm proximal of the anus. Normal saline, acetic acid, crocin (5.00, 10.00 and 20.00 mg kg-1), mesalazine (100 and 300 mg kg-1) and crocin (5.00 mg kg-1) plus mesalazine (100 mg kg-1) were administered after induction of colitis for eight days. Body weight, organosomatic index (OSI), macroscopic and microscopic evaluations of colon and measurement of malondialdehyde (MDA), superoxide dismutase (SOD), tumor necrosis factor-alpha (TNF-α) contents of colon tissue were determined on day eight after induction of colitis. Crocin (10.00 and 20.00 mg kg-1), mesalazine (300 mg kg-1) and crocin (5.00 mg kg-1) plus mesalazine (100 mg kg-1) significantly (p < 0.05) improved body weight and OSI and reduced macroscopic and microscopic scores. These treatments also significantly (p <0.05) recovered the increased levels of MDA and TNF-α as well as the decreased level of SOD in colon tissue. Crocin and mesalazine did not produce significant effects in intact rats. Based on the results, it is concluded that crocin and mesalazine produced protective effects on colon tissue via antioxidant and anti-inflammatory actions. In addition, a synergistic effect was observed between crocin and mesalazine in attenuating ulcerative colitis.
Sigma-Aldrich
Crocin