MilliporeSigma
  • Cooperative Blockade of PKCα and JAK2 Drives Apoptosis in Glioblastoma.
Cooperative Blockade of PKCα and JAK2 Drives Apoptosis in Glioblastoma.
Cancer research (2019-12-07)
Robyn A Wong, Xujun Luo, Mimi Lu, Zhenyi An, Daphne A Haas-Kogan, Joanna J Phillips, Kevan M Shokat, William A Weiss, Qi Wen Fan
ABSTRACT
The mTOR signaling is dysregulated prominently in human cancers including glioblastoma, suggesting mTOR as a robust target for therapy. Inhibitors of mTOR have had limited success clinically, however, in part because their mechanism of action is cytostatic rather than cytotoxic. Here, we tested three distinct mTOR kinase inhibitors (TORKi) PP242, KU-0063794, and sapanisertib against glioblastoma cells. All agents similarly decreased proliferation of glioblastoma cells, whereas PP242 uniquely induced apoptosis. Apoptosis induced by PP242 resulted from off-target cooperative inhibition of JAK2 and protein kinase C alpha (PKCα). Induction of apoptosis was also decreased by additional on-target inhibition of mTOR, due to induction of autophagy. As EGFR inhibitors can block PKCα, EGFR inhibitors erlotinib and osimertinib were tested separately in combination with the JAK2 inhibitor AZD1480. Combination therapy induced apoptosis of glioblastoma tumors in both flank and in patient-derived orthotopic xenograft models, providing a preclinical rationale to test analogous combinations in patients. SIGNIFICANCE: These findings identify PKCα and JAK2 as targets that drive apoptosis in glioblastoma, potentially representing a clinically translatable approach for glioblastoma.
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone 4G10®, clone 4G10®, Upstate®, from mouse
Sigma-Aldrich
MISSION® esiRNA, targeting human JAK2
Sigma-Aldrich
Anti-GAPDH Antibody, from chicken, purified by affinity chromatography