MilliporeSigma
  • An Nd3+-Sensitized Upconversion Fluorescent Sensor for Epirubicin Detection.
An Nd3+-Sensitized Upconversion Fluorescent Sensor for Epirubicin Detection.
Nanomaterials (Basel, Switzerland) (2019-12-05)
Jingwen Mo, Long Shen, Qian Xu, Jiaying Zeng, Jingjie Sha, Tao Hu, Kedong Bi, Yunfei Chen
ABSTRACT
We describe here an Nd3+-sensitized upconversion fluorescent sensor for epirubicin (EPI) detection in aqueous solutions under 808 nm laser excitation. The upconversion fluorescence of nanoparticles is effectively quenched in the presence of EPI via a fluorescence resonance energy transfer mechanism. The dynamic quenching constant was 2.10 × 104 M-1. Normalized fluorescence intensity increased linearly as the EPI concentration was raised from 0.09 μM to 189.66 μM and the fluorometric detection limit was 0.05 μM. The sensing method was simple, fast, and low-cost and was able to be applied to determine the levels of EPI in urine with spike recoveries from 97.5% to 102.6%. Another important feature of the proposed fluorescent sensor is that it holds a promising potential for in vivo imaging and detection due to its distinctive properties such as weak autofluorescence, low heating effect, and high light penetration depth.
Sigma-Aldrich
Epirubicin hydrochloride, ≥90% (HPLC)
Sigma-Aldrich
Neodymium(III) acetate hydrate, 99.9%
Sigma-Aldrich
Ytterbium(III) acetate hydrate, 99.95% trace metals basis
Sigma-Aldrich
Erbium(III) acetate hydrate, 99.9% trace metals basis
Sigma-Aldrich
Yttrium(III) acetate hydrate, 99.9% metals basis