MilliporeSigma
  • Lymphoblast-derived hiPS cell lines generated from four individuals of a family of genetically unrelated parents and their female monozygotic twins.
Lymphoblast-derived hiPS cell lines generated from four individuals of a family of genetically unrelated parents and their female monozygotic twins.
Stem cell research (2019-11-18)
Marga J Bouma, Christian Freund, Adriaan P IJzerman, Dorret I Boomsma, Christine L Mummery, Karine Raymond
ABSTRACT
Lymphoblast cells from four individuals of a family of two genetically unrelated parents and their monozygotic twins were used to generate integration-free human induced pluripotent stem cells (hiPSCs). Reprogramming factors were delivered by co-electroporation of three episomal-based plasmids expressing OCT3/4, SOX2, KLF4, L-MYC and LIN28. The hiPSCs showed a normal karyotype, expressed pluripotency-associated markers, displayed the capacity for in vitro differentiation into the three germ layers and were Epstein Barr virus-free. These hiPSC lines offer the possibility to compare genetically unrelated and genetically identical tissues from different individuals and to study genotype-specific effects, which are particularly relevant for toxicology testing.
Sigma-Aldrich
RPMI-1640 Medium, HEPES Modification, With 25 mM HEPES, without L-glutamine., liquid, sterile-filtered, suitable for cell culture