MilliporeSigma
  • A New Animal Model for Investigation of Mechanical Unloading in Hypertrophic and Failing Hearts: Combination of Transverse Aortic Constriction and Heterotopic Heart Transplantation.
A New Animal Model for Investigation of Mechanical Unloading in Hypertrophic and Failing Hearts: Combination of Transverse Aortic Constriction and Heterotopic Heart Transplantation.
PloS one (2016-02-04)
Andreas Schaefer, Yvonne Schneeberger, Justus Stenzig, Daniel Biermann, Marisa Jelinek, Hermann Reichenspurner, Thomas Eschenhagen, Heimo Ehmke, Alexander P Schwoerer
ABSTRACT
Previous small animal models for simulation of mechanical unloading are solely performed in healthy or infarcted hearts, not representing the pathophysiology of hypertrophic and dilated hearts emerging in heart failure patients. In this article, we present a new and economic small animal model to investigate mechanical unloading in hypertrophic and failing hearts: the combination of transverse aortic constriction (TAC) and heterotopic heart transplantation (hHTx) in rats. To induce cardiac hypertrophy and failure in rat hearts, three-week old rats underwent TAC procedure. Three and six weeks after TAC, hHTx with hypertrophic and failing hearts in Lewis rats was performed to induce mechanical unloading. After 14 days of mechanical unloading animals were euthanatized and grafts were explanted for further investigations. 50 TAC procedures were performed with a survival of 92% (46/50). When compared to healthy rats left ventricular surface decreased to 5.8±1.0 mm² (vs. 9.6± 2.4 mm²) (p = 0.001) after three weeks with a fractional shortening (FS) of 23.7± 4.3% vs. 28.2± 1.5% (p = 0.01). Six weeks later, systolic function decreased to 17.1± 3.2% vs. 28.2± 1.5% (p = 0.0001) and left ventricular inner surface increased to 19.9±1.1 mm² (p = 0.0001). Intraoperative graft survival during hHTx was 80% with 46 performed procedures (37/46). All transplanted organs survived two weeks of mechanical unloading. Combination of TAC and hHTx in rats offers an economic and reproducible small animal model enabling serial examination of mechanical unloading in a truly hypertrophic and failing heart, representing the typical pressure overloaded and dilated LV, occurring in patients with moderate to severe heart failure.
Sigma-Aldrich
Anti-Dystrophin Antibody, clone 1808, clone 1808, Chemicon®, from mouse