MilliporeSigma
  • Uncoupling protein 2 impacts endothelial phenotype via p53-mediated control of mitochondrial dynamics.
Uncoupling protein 2 impacts endothelial phenotype via p53-mediated control of mitochondrial dynamics.
Circulation research (2013-07-04)
Yukio Shimasaki, Ning Pan, Louis M Messina, Chunying Li, Kai Chen, Lijun Liu, Marcus P Cooper, Joseph A Vita, John F Keaney
ABSTRACT
Mitochondria, although required for cellular ATP production, are also known to have other important functions that may include modulating cellular responses to environmental stimuli. However, the mechanisms whereby mitochondria impact cellular phenotype are not yet clear. To determine how mitochondria impact endothelial cell function. We report here that stimuli for endothelial cell proliferation evoke strong upregulation of mitochondrial uncoupling protein 2 (UCP2). Analysis in silico indicated increased UCP2 expression is common in highly proliferative cell types, including cancer cells. Upregulation of UCP2 was critical for controlling mitochondrial membrane potential (Δψ) and superoxide production. In the absence of UCP2, endothelial growth stimulation provoked mitochondrial network fragmentation and premature senescence via a mechanism involving superoxide-mediated p53 activation. Mitochondrial network fragmentation was both necessary and sufficient for the impact of UCP2 on endothelial cell phenotype. These data identify a novel mechanism whereby mitochondria preserve normal network integrity and impact cell phenotype via dynamic regulation of UCP2.