MilliporeSigma
  • Stimulation of astrocyte fatty acid oxidation by thyroid hormone is protective against ischemic stroke-induced damage.
Stimulation of astrocyte fatty acid oxidation by thyroid hormone is protective against ischemic stroke-induced damage.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism (2016-02-14)
Naomi L Sayre, Mikaela Sifuentes, Deborah Holstein, Sheue-Yann Cheng, Xuguang Zhu, James D Lechleiter
ABSTRACT
We previously demonstrated that stimulation of astrocyte mitochondrial ATP production via P2Y1 receptor agonists was neuroprotective after cerebral ischemic stroke. Another mechanism that increases ATP production is fatty acid oxidation (FAO). We show that in primary human astrocytes, FAO and ATP production are stimulated by 3,3,5 triiodo-l-thyronine (T3). We tested whether T3-stimulated FAO enhances neuroprotection, and show that T3 increased astrocyte survival after either hydrogen peroxide exposure or oxygen glucose deprivation. T3-mediated ATP production and protection were both eliminated with etomoxir, an inhibitor of FAO. T3-mediated protection in vitro was also dependent on astrocytes expressing HADHA (hydroxyacyl-CoA dehydrogenase/3-ketoacyl-CoA thiolase/enoyl-CoA hydratase), which we previously showed was critical for T3-mediated FAO in fibroblasts. Consistent with previous reports, T3-treatment decreased stroke volumes in mice. While T3 decreased stroke volume in etomoxir-treated mice, T3 had no protective effect on stroke volume in HADHA +/- mice or in mice unable to upregulate astrocyte-specific energy production. In vivo, 95% of HADHA co-localize with glial-fibrillary acidic protein, suggesting the effect of HADHA is astrocyte mediated. These results suggest that astrocyte-FAO modulates lesion size and is required for T3-mediated neuroprotection post-stroke. To our knowledge, this is the first report of a neuroprotective role for FAO in the brain.
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein Antibody, Chemicon®, from chicken
Sigma-Aldrich
Anti-O4 Antibody, clone 81, clone 81 (mAB O4), Chemicon®, from mouse
Sigma-Aldrich
Anti-Oligodendrocytes Antibody, clone NS-1, ascites fluid, clone NS-1, Chemicon®