MilliporeSigma
  • The Effects of Anti-Vascular Endothelial Growth Factor Drugs on Retinal Pigment Epithelial Cell Culture.
The Effects of Anti-Vascular Endothelial Growth Factor Drugs on Retinal Pigment Epithelial Cell Culture.
Turkish journal of ophthalmology (2018-09-12)
Mustafa Şahiner, Dilek Bahar, Ayşe Öner, Zeynep Burçin Gönen, Metin Ünlü, Duygu Gülmez Sevim, Çağatay Karaca, Galip Ertuğrul Mirza
ABSTRACT
To assess the effects of anti-vascular endothelial growth factor (VEGF) drugs on retinal pigment epithelium cell culture. Aflibercept (0.5 mg/mL), bevacizumab (0.3125 mg/mL), and ranibizumab (0.125 mg/mL) were applied to retinal pigment epithelium cell cultures isolated from the enucleated eyes of New Zealand white rabbits. Viability, apoptosis, proliferation, and senescence of the cells were evaluated in control and drug-treated cultures at the end of 72 hours. Cells treated with aflibercept showed increased viability and decreased apoptosis compared to the control culture and both the bevacizumab- and ranibizumab-treated groups (p<0.05). Statistically increased apoptosis and decreased viability were found in the bevacizumab and ranibizumab-treated groups compared with the control group (p<0.05). There were no statistically significant differences in cell proliferation and senescence between the groups (p>0.05). Anti-VEGF drugs did not affect senescence or proliferation of retinal pigment epithelium cells. Aflibercept was found to decrease apoptosis and increase cell viability, while ranibizumab and bevacizumab increased apoptosis and reduced cell viability in retinal pigment epithelium culture.
Sigma-Aldrich
Anti-Cytokeratin 18 Antibody, clone RGE53, clone RGE53, Chemicon®, from mouse