MilliporeSigma
  • Aβ induces astrocytic glutamate release, extrasynaptic NMDA receptor activation, and synaptic loss.
Aβ induces astrocytic glutamate release, extrasynaptic NMDA receptor activation, and synaptic loss.
Proceedings of the National Academy of Sciences of the United States of America (2013-06-19)
Maria Talantova, Sara Sanz-Blasco, Xiaofei Zhang, Peng Xia, Mohd Waseem Akhtar, Shu-ichi Okamoto, Gustavo Dziewczapolski, Tomohiro Nakamura, Gang Cao, Alexander E Pratt, Yeon-Joo Kang, Shichun Tu, Elena Molokanova, Scott R McKercher, Samuel Andrew Hires, Hagit Sason, David G Stouffer, Matthew W Buczynski, James P Solomon, Sarah Michael, Evan T Powers, Jeffery W Kelly, Amanda Roberts, Gary Tong, Traci Fang-Newmeyer, James Parker, Emily A Holland, Dongxian Zhang, Nobuki Nakanishi, H-S Vincent Chen, Herman Wolosker, Yuqiang Wang, Loren H Parsons, Rajesh Ambasudhan, Eliezer Masliah, Stephen F Heinemann, Juan C Piña-Crespo, Stuart A Lipton
ABSTRACT
Synaptic loss is the cardinal feature linking neuropathology to cognitive decline in Alzheimer's disease (AD). However, the mechanism of synaptic damage remains incompletely understood. Here, using FRET-based glutamate sensor imaging, we show that amyloid-β peptide (Aβ) engages α7 nicotinic acetylcholine receptors to induce release of astrocytic glutamate, which in turn activates extrasynaptic NMDA receptors (eNMDARs) on neurons. In hippocampal autapses, this eNMDAR activity is followed by reduction in evoked and miniature excitatory postsynaptic currents (mEPSCs). Decreased mEPSC frequency may reflect early synaptic injury because of concurrent eNMDAR-mediated NO production, tau phosphorylation, and caspase-3 activation, each of which is implicated in spine loss. In hippocampal slices, oligomeric Aβ induces eNMDAR-mediated synaptic depression. In AD-transgenic mice compared with wild type, whole-cell recordings revealed excessive tonic eNMDAR activity accompanied by eNMDAR-sensitive loss of mEPSCs. Importantly, the improved NMDAR antagonist NitroMemantine, which selectively inhibits extrasynaptic over physiological synaptic NMDAR activity, protects synapses from Aβ-induced damage both in vitro and in vivo.
Sigma-Aldrich
NitroSynapsin hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Anti-Tau Antibody, a.a. 210-241, clone Tau-5, ascites fluid, clone Tau-5, Chemicon®