MilliporeSigma
  • Long-term treatment with testosterone alters ovary innervation in adult pigs.
Long-term treatment with testosterone alters ovary innervation in adult pigs.
Journal of ovarian research (2016-10-12)
Barbara Jana, Karolina A Meller, Michał Bulc, Jarosław Całka
ABSTRACT
Intraovarian distribution and density of nerve fibres immunoreactive (IR) to protein gene product 9.5 (PGP 9.5) and containing dopamine-β-hydroxylase (DβH), neuropeptide Y (NPY), somatostatin (SOM), galanin (GAL) were determined. From day 4 of the first oestrous cycle to day 20 of the second studied cycle, experimental gilts (n = 3) were injected with testosterone (T), while control gilts (n = 3) received corn oil. After T administration the numbers of fibres IR to PGP 9.5 and fibres IR to DβH, NPY and SOM were decreased. Fewer PGP 9.5- and DβH-IR terminals were observed within the ground plexus and around arteries and medullar veins, and medium tertiary follicles, and DβH-IR terminals in the vicinity of small tertiary follicles. T decreased the density of NPY-IR fibres in the medullar part of the ground plexus, and SOM-IR in the cortical part of the ground plexus. The obtained data show that long-term T treatment of gilts decreases the total number of intraovarian fibres, including sympathetic ones. These results suggest that elevated T levels that occur during pathological states may affect the innervation pattern of ovaries, and their function(s).
Sigma-Aldrich
Anti-Dopamine β Hydroxylase Antibody, clone 4F10.2, ascites fluid, clone 4F10.2, Chemicon®
Sigma-Aldrich
Anti-Galanin Antibody, serum, from rabbit
Sigma-Aldrich
Anti-Dopamine β Hydroxylase Antibody, serum, Chemicon®