MilliporeSigma
  • The expression and the regulatory role of OX40 and 4-1BB heterodimer in activated human T cells.
The expression and the regulatory role of OX40 and 4-1BB heterodimer in activated human T cells.
Blood (2005-06-09)
Bruce Y Ma, Sebastian A Mikolajczak, Ali Danesh, Karoline A Hosiawa, Cheryl M Cameron, Akifumi Takaori-Kondo, Takashi Uchiyama, David J Kelvin, Atsuo Ochi
ABSTRACT
OX40 and 4-1BB are members of the tumor necrosis factor (TNF) family of costimulatory receptors whose signaling is important for differential immune responses mediated by CD4+ or CD8+ T cells. Although activated T cells may acquire OX40/4-1BB double-positive phenotype and signaling from each receptor is expected to influence cell functions, the relevance between OX40 and 4-1BB has never been investigated before. While we were investigating the expression of OX40 and 4-1BB on activated human T cells, we found that they colocalize. The study of receptor gene-transfected cells showed that both receptors coendocytose and the complex of OX40 and 4-1BB was detected by specific ligands or antibodies (Abs). The heterodimer of OX40 and 4-1BB was identified by sodium dodecyl sulfate-polyacrylamide gel electrophoresis (SDS-PAGE) under nonreduced conditions and was associated with the tumor receptor-associated factor (TRAF) family proteins in a unique manner. Furthermore, the stimulation of OX40/4-1BB rendered cells sensitive to apoptosis induced by TNF-alpha that accompanied reduced activation of nuclear factor-kappaB (NF-kappaB). Finally, the OX40/4-1BB stimulation repressed the mitogen response in activated CD25+CD4+ T cells and preactivated CD8+ T cells. Thus, the OX40/4-1BB heterodimer appears to represent a unique regulatory receptor in activated T cells.
Sigma-Aldrich
Anti-DYKDDDDK (FLAG® epitope tag) Antibody, clone 2EL-1B11, ascites fluid, clone 2EL-1B11, Chemicon®