MilliporeSigma
  • A Transcription Factor Pulse Can Prime Chromatin for Heritable Transcriptional Memory.
A Transcription Factor Pulse Can Prime Chromatin for Heritable Transcriptional Memory.
Molecular and cellular biology (2016-12-07)
Aimee Iberg-Badeaux, Samuel Collombet, Benoit Laurent, Chris van Oevelen, Kuo-Kai Chin, Denis Thieffry, Thomas Graf, Yang Shi
ABSTRACT
Short-term and long-term transcriptional memory is the phenomenon whereby the kinetics or magnitude of gene induction is enhanced following a prior induction period. Short-term memory persists within one cell generation or in postmitotic cells, while long-term memory can survive multiple rounds of cell division. We have developed a tissue culture model to study the epigenetic basis for long-term transcriptional memory (LTTM) and subsequently used this model to better understand the epigenetic mechanisms that enable heritable memory of temporary stimuli. We find that a pulse of transcription factor CCAAT/enhancer-binding protein alpha (C/EBPα) induces LTTM on a subset of target genes that survives nine cell divisions. The chromatin landscape at genes that acquire LTTM is more repressed than at those genes that do not exhibit memory, akin to a latent state. We show through chromatin immunoprecipitation (ChIP) and chemical inhibitor studies that RNA polymerase II (Pol II) elongation is important for establishing memory in this model but that Pol II itself is not retained as part of the memory mechanism. More generally, our work reveals that a transcription factor involved in lineage specification can induce LTTM and that failure to rerepress chromatin is one epigenetic mechanism underlying transcriptional memory.
Sigma-Aldrich
Anti-Trimethyl Histone H3 (Lys4) Antibody, clone CMA304, clone CMA304, from mouse
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys27) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-dimethyl-Histone H3 (Lys4) Antibody, clone AW30, rabbit monoclonal, culture supernatant, clone AW30, Upstate®