MilliporeSigma
  • The Dysfunctional MDM2-p53 Axis in Adipocytes Contributes to Aging-Related Metabolic Complications by Induction of Lipodystrophy.
The Dysfunctional MDM2-p53 Axis in Adipocytes Contributes to Aging-Related Metabolic Complications by Induction of Lipodystrophy.
Diabetes (2018-08-23)
Zhuohao Liu, Leigang Jin, Jin-Kui Yang, Baile Wang, Kelvin K L Wu, Philip Hallenborg, Aimin Xu, Kenneth K Y Cheng
ABSTRACT
Profound loss and senescence of adipose tissues are hallmarks of advanced age, but the underlying cause and their metabolic consequences remain obscure. Proper function of the murine double minute 2 (MDM2)-p53 axis is known to prevent tumorigenesis and several metabolic diseases, yet its role in regulation of adipose tissue aging is still poorly understood. In this study, we show that the proximal p53 inhibitor MDM2 is markedly downregulated in subcutaneous white and brown adipose tissues of mice during aging. Genetic disruption of MDM2 in adipocytes triggers canonical p53-mediated apoptotic and senescent programs, leading to age-dependent lipodystrophy and its associated metabolic disorders, including type 2 diabetes, nonalcoholic fatty liver disease, hyperlipidemia, and energy imbalance. Surprisingly, this lipodystrophy mouse model also displays premature loss of physiological integrity, including impaired exercise capacity, multiple organ senescence, and shorter life span. Transplantation of subcutaneous fat rejuvenates the metabolic health of this aging-like lipodystrophy mouse model. Furthermore, senescence-associated secretory factors from MDM2-null adipocytes impede adipocyte progenitor differentiation via a non-cell-autonomous manner. Our findings suggest that tight regulation of the MDM2-p53 axis in adipocytes is required for adipose tissue dynamics and metabolic health during the aging process.
Sigma-Aldrich
Oil Red O, certified by the BSC
Sigma-Aldrich
Anti-MDM2 Antibody, clone 3G9, clone 3G9, from mouse
Roche
In Situ Cell Death Detection Kit, Fluorescein, sufficient for ≤50 tests, suitable for detection