MilliporeSigma
  • The Effects of Nonporous Silica Nanoparticles on Cultured Human Keratocytes.
The Effects of Nonporous Silica Nanoparticles on Cultured Human Keratocytes.
Investigative ophthalmology & visual science (2017-01-25)
Bora Yim, Joo-Hee Park, Hyejoong Jeong, Jinkee Hong, Young-Joo Shin, Roy S Chuck, Choul Yong Park
ABSTRACT
Silica nanoparticles (SiNPs) are promising carriers for ophthalmic drug delivery. In this study, we investigated the effect of various sizes of nonporous SiNPs on cultured human keratocytes. Three different sizes of SiNPs (50, 100, and 150 nm) were manufactured. Primarily cultured human keratocytes were exposed to different concentrations (0, 25, 50, and 100 μg/mL) of three sizes of SiNPs for up to 72 hours. Intracellular reactive oxygen species (ROS) generation, cellular viability, lactate dehydrogenase (LDH) assay, autophagy, vimentin expression, and mammalian target of rapamycin (mTOR) pathway activation were evaluated. Intracellular distribution of SiNPs was evaluated with transmission electron microscopy. Transmission electron microscopy revealed SiNPs were taken up by keratocytes inside cytoplasmic vacuoles. Neither nuclear entry of SiNPs nor mitochondrial structural damage was observed. Both intracellular ROS generation and LDH level remained unchanged with up to 100 μg/mL SiNP treatment. Cellular viability was not affected by SiNP treatment. Autophagy showed significant dose-dependent activation with 50- and 100-nm SiNPs. However, mTOR activation remained unchanged. Vimentin expression did not show any significant increase with SiNPs. Our findings suggested that 50-, 100-, and 150-nm SiNPs did not induce significant cytotoxicity in cultured human keratocytes at concentrations up to 100 μg/mL.
Sigma-Aldrich
Anti-Vimentin Antibody, clone V9, clone V9, Chemicon®, from mouse
Sigma-Aldrich
Fluorometric Intracellular Ros Kit, sufficient for 200 fluorometric tests (orange)