MilliporeSigma
  • Impact of defined matrix interactions on insulin production by cultured human beta-cells: effect on insulin content, secretion, and gene transcription.
Impact of defined matrix interactions on insulin production by cultured human beta-cells: effect on insulin content, secretion, and gene transcription.
Diabetes (2006-09-28)
Thomas Kaido, Mayra Yebra, Vincenzo Cirulli, Christopher Rhodes, Giuseppe Diaferia, Anthony M Montgomery
ABSTRACT
The impact of extracellular matrix on insulin production needs to be understood both to optimize the derivation of functional beta-cells for transplantation and to understand mechanisms controlling islet neogenesis and glucose homeostasis. In this study, we present evidence that adhesion to some common matrix constituents has a profound impact on the transcription, secretion, and storage of insulin by human beta-cells. The integrin-dependent adhesion of fetal beta-cells to both collagen IV and vitronectin induces significant glucose-independent insulin secretion and a substantial reciprocal decline in insulin content. Collagen IV, but not vitronectin, induces comparable responses in adult beta-cells. Inhibition of extracellular signal-regulated kinase activation abrogates matrix-induced insulin secretion and effectively preserves the insulin content of adherent beta-cells. Using real-time PCR, we demonstrate that adhesion of both fetal and adult beta-cells to collagen IV and vitronectin also results in the marked suppression of insulin gene transcription. Based on these findings, we contend that integrin-dependent adhesion and signaling in response to certain matrices can have a significant negative impact on insulin production by primary human beta-cells. Such responses were not found to be associated with cell death but may precede beta-cell dedifferentiation.
Sigma-Aldrich
Anti-Integrin αVβ3 Antibody, clone LM609, clone LM609, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin β1 Antibody, clone P4C10, azide free, clone P4C10, Chemicon®, from mouse