MilliporeSigma
  • Improving Cell Survival in Injected Embryos Allows Primed Pluripotent Stem Cells to Generate Chimeric Cynomolgus Monkeys.
Improving Cell Survival in Injected Embryos Allows Primed Pluripotent Stem Cells to Generate Chimeric Cynomolgus Monkeys.
Cell reports (2018-11-30)
Yu Kang, Zongyong Ai, Kui Duan, Chenyang Si, Yong Wang, Yun Zheng, Jingjing He, Yu Yin, Shumei Zhao, Baohua Niu, Xiaoqing Zhu, Li Liu, Lifeng Xiang, Linming Zhang, Yuyu Niu, Weizhi Ji, Tianqing Li
ABSTRACT
Monkeys are an optimal model species for developing stem cell therapies. We previously reported generating chimeric cynomolgus monkey fetuses using dome-shaped embryonic stem cells (dESCs). However, conventional primed pluripotent stem cells (pPSCs) lack chimera competency. Here, by altering the media in which injected morulae are cultured, we observed increased survival of cynomolgus monkey primed ESCs, induced PSCs, and somatic cell nuclear transfer-derived ESCs, thereby enabling chimeric contributions with 0.1%-4.5% chimerism into the embryonic and placental tissues, including germ cell progenitors in chimeric monkeys. Mechanically, dESCs and pPSCs belong to different cell types and similarly express epiblast ontogenic genes. The host embryonic microenvironment could reprogram injected PSCs to embryonic-like cells. However, the reprogramming level and chimerism were associated with the cell state of injected PSCs. Our findings provide a method to understand pluripotency and broaden the use of embryonic chimeras for basic developmental biology research and regenerative medicine.
Sigma-Aldrich
Anti-Sox2 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
ES Cell Characterization Kit, The Embryonic Stem Cell Characterization Kit phenotypically assesses the differentiation status of ES cells by measuring their AP activity, cell-surface stage-specific antigens (SSEA-1, SSEA-4) as well as expression of TRA-1-60, TRA-1-81 antigens.
Sigma-Aldrich
Anti-α-Actinin (Sarcomeric) antibody, Mouse monoclonal, clone EA-53, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Tubulin Antibody, beta III isoform, CT, clone TU-20 (Similar to TUJ1), ascites fluid, clone TU-20 (Similar to TUJ1), Chemicon®
Sigma-Aldrich
Anti-PRDM14 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-Nestin Antibody, clone 10C2, clone 10C2, Chemicon®, from mouse
Sigma-Aldrich
Anti-NeuN Antibody (rabbit), from rabbit, purified by affinity chromatography