MilliporeSigma
Expression of NADPH oxidase in human pancreatic islets.
Life sciences (2012-07-24)
Eduardo Rebelato, Thiago R Mares-Guia, Maria Fernanda R Graciano, Letícia Labriola, Luiz R G Britto, Humberto M Garay-Malpartida, Rui Curi, Mari C Sogayar, Angelo R Carpinelli
ABSTRACT
NADPH oxidase (NOX) is a known source of superoxide anions in phagocytic and non-phagocytic cells. In this study, the presence of this enzyme in human pancreatic islets and the importance of NADPH oxidase in human β-cell function were investigated. In isolated human pancreatic islets, the expression of NADPH oxidase components was evidenced by real-time PCR (p22(PHOX), p47(PHOX) and p67(PHOX)), Western blotting (p47(PHOX) and p67(PHOX)) and immunohistochemistry (p47(PHOX), p67(PHOX) and gp91(PHOX)). Immunohistochemistry experiments showed co-localization of p47(PHOX), p67(PHOX) and gp91(PHOX) (isoform 2 of NADPH oxidase-NOX2) with insulin secreting cells. Inhibition of NADPH oxidase activity impaired glucose metabolism and glucose-stimulated insulin secretion. These findings demonstrate the presence of the main intrinsic components of NADPH oxidase comprising the NOX2 isoform in human pancreatic islets, whose activity also contributes to human β-cell function.
Sigma-Aldrich
Anti-p47-phox Antibody, serum, Upstate®