MilliporeSigma
  • Renal expression of SIBLING proteins and their partner matrix metalloproteinases (MMPs).
Renal expression of SIBLING proteins and their partner matrix metalloproteinases (MMPs).
Kidney international (2005-06-16)
Kalu U E Ogbureke, Larry W Fisher
ABSTRACT
Three members of the small integrin-binding ligand N-linked glycoprotein (SIBLING) family of proteins have recently been shown to bind and activate specific promatrix metalloproteinases (MMPs) and to overcome the inhibition of tissue inhibitors of MMPs (TIMPs). Although usually associated with mineralized tissues, we have shown that the SIBLINGs and their MMP partners, when known, are coexpressed in salivary gland ductal cells. The present study examined the expression patterns of both the SIBLINGs and their MMP partners in adult kidney. The expression patterns of all five SIBLINGs known to date, and their MMP partners were determined in monkey kidney using immunohistochemistry and in situ hybridization techniques. Bone sialoprotein (BSP) and its partner, MMP-2, were coexpressed in both the proximal and distal tubules. Osteopontin, as previously shown, was expressed in the distal tubules while its partner MMP-3 was expressed in both the proximal tubule and distal tubles. Dentin matrix protein-1 (DMP1) and MMP-9 were coexpressed throughout the nephron, including both parietal cells of Bowman's capsule and the thin limb of the loop of Henle. Dentin sialophosphoprotein (DSPP) and matrix extracellular phosphoglycoprotein (MEPE) were expressed in the proximal tubule and distal tubule, and proximal tubule, respectively. In contrast to salivary gland in which all SIBLINGs and their MMP partners were coexpressed throughout the length of the ducts, these proteins were differentially expressed within the normal adult nephron. We hypothesize that the cells use the SIBLING/MMP pairs in the normal turnover of cell surface proteins and/or pericellular matrix proteins such as those in basement membranes.
Sigma-Aldrich
Anti-MMP-3 Antibody, cross reacts w/hMMP-10, clone 55-2A4, clone 55-2A4, Chemicon®, from mouse