MilliporeSigma
  • Increased expression levels of integrin α9β1 and CD11b on circulating neutrophils and elevated serum IL-17A in elderly aspiration pneumonia.
Increased expression levels of integrin α9β1 and CD11b on circulating neutrophils and elevated serum IL-17A in elderly aspiration pneumonia.
Respiration; international review of thoracic diseases (2012-12-22)
Yasuyuki Taooka, Miki Ohe, Lijun Chen, Akihisa Sutani, Yusuke Higashi, Takeshi Isobe
ABSTRACT
Repeated aspiration pneumonia is a serious problem in the elderly. In aspiration pneumonia, neutrophils play an important role in acute lung injury, while CD18-independent neutrophil transmigration pathways have also been reported in acid-aspiration pneumonia animal models. However, the involvement of IL-17A and β1 integrin still remains unclear. The β1 integrin subfamily integrin α9β1 has been shown to be expressed on human neutrophils and to mediate adhesion to extracellular matrix proteins including the vascular cell adhesion molecule-1. To elucidate the possible involvement of β1 integrin subfamily and IL-17A in aspiration pneumonia. We analyzed the expression levels of CD11b, CD18 and integrin α9β1 in circulating neutrophils and serum concentration of IL-17A, IL-22 and IL-23 in elderly aspiration pneumonia patients (n = 32, 14 males and 18 females, 78.8 ± 3.9 years old) at 2 time points (on the day of admission before starting antibiotics and the day after finishing antibiotics) and compared the results with those of a control group (n = 30, 13 males and 17 females, 76.1 ± 3.4 years old). Recombinant IL-17A stimulated integrin α9β1 and CD11b expression levels in healthy human neutrophils in vitro. The expression levels of integrin α9β1 and CD11b in circulating neutrophils were significantly higher in pneumonia patients compared with the controls. In addition, serum IL-17A concentration was significantly increased in pneumonia patients. Integrin α9β1 levels positively correlated with serum IL-17A and CD18 expression levels. These findings suggest a potential role of integrin α9β1 expressed in neutrophils and elevated serum IL-17A in extravasation of neutrophils in cases of aspiration pneumonia.
Sigma-Aldrich
Anti-Integrin β6 Antibody, clone R6G9, azide free, clone R6G9, Chemicon®, from mouse