MilliporeSigma
  • Integrin mechanotransduction stimulates caveolin-1 phosphorylation and recruitment of Csk to mediate actin reorganization.
Integrin mechanotransduction stimulates caveolin-1 phosphorylation and recruitment of Csk to mediate actin reorganization.
American journal of physiology. Heart and circulatory physiology (2004-10-09)
C Radel, V Rizzo
ABSTRACT
To identify the role of caveolin-1 in integrin mechanotransduction, we exposed bovine aortic endothelial cells to 10 dyn/cm2 of laminar shear stress. Caveolin-1 was acutely and transiently phosphorylated with shear, occurring downstream of beta1-integrin activation as the beta1-integrin blocking antibody JB1A was inhibitory. In manipulating Src family kinase (SFK) activity with knockdown of Csk or type 1 protein phosphatase (PP1) treatment, we observed coordinate increase and decrease in shear-induced caveolin-1 phosphorylation, respectively. Hence, shear-stimulated caveolin-1 phosphorylation is regulated by SFKs. Shear-induced recruitment and phosphorylation of caveolin-1 occurred at beta1-integrin sites in a beta1-integrin- and SFK-dependent manner. Csk, described to interact with pY14-caveolin-1 and integrins, bound to an increased pool of phosphorylated caveolin-1 after shear corresponding with elevated Csk at beta1-integrin sites. Like caveolin-1, treatment with JB1A and PP1 attenuated shear-induced Csk association with beta1-integrins. Csk function was assayed with transfection of a caveolin-1 phosphorylation domain peptide. The peptide attenuated shear-induced association of Csk at beta1-integrin sites, as well as colocalization of Csk with paxillin and phosphorylated caveolin-1. Because integrin and Csk activity regulate cytoskeletal reorganization, we evaluated the role of this mechanism in shear-induced myosin light chain (MLC) phosphorylation. Knockdown of Csk expression was sufficient to reduce MLC diphosphorylation due to shear. Disruption of Csk-integrin association by peptide treatment was also inhibitory of the MLC diphosphorylation response. Together these data indicate that integrin activation with shear stress results in SFK-regulated caveolin-1 phosphorylation that, in turn, mediates Csk association at integrin sites, where it plays a role in downstream, shear-stimulated MLC diphosphorylation.
Sigma-Aldrich
Anti-Integrin β1 Antibody, a.a. 82-87, clone JB1A (a.k.a. J10), ascites fluid, clone JB1A (J10), Chemicon®
Sigma-Aldrich
Anti-Integrin β1 Antibody, clone P4G11, clone P4G11, Chemicon®, from mouse