MilliporeSigma
  • Human airway trypsin-like protease increases mucin gene expression in airway epithelial cells.
Human airway trypsin-like protease increases mucin gene expression in airway epithelial cells.
American journal of respiratory cell and molecular biology (2003-09-23)
Manabu Chokki, Satoshi Yamamura, Hiroshi Eguchi, Tsukio Masegi, Hideki Horiuchi, Hirofumi Tanabe, Takashi Kamimura, Susumu Yasuoka
ABSTRACT
Human airway trypsin-like protease (HAT) is a serine protease found in sputum of patients with chronic airway diseases and is an agonist of protease-activated receptor-2 (PAR-2). Results from this study show that HAT treatment also enhances mucus production by the airway epithelial cell line NCI-H292 in vitro. Histologic examination showed that HAT enhances mucous glycoconjugate synthesis, whereas the PAR-2 agonist peptide (PAR-2 AP) has no such effect. HAT, but not PAR-2 AP, enhances MUC2 and MUC5AC gene expression 23-fold and 32-fold, respectively. The proteolytic activity of HAT is required to enhance MUC5AC gene expression; the addition of the inhibitors of trypsin-like protease activity of HAT, aprotinin and leupeptin, abolishes its enhancing effect. AG1478, anti-epidermal growth factor receptor (anti-EGFR)-neutralizing antibody, and anti-amphiregulin (AR)-neutralizing antibody all inhibited the stimulatory effect of HAT. Furthermore, HAT increases AR gene expression and subsequent AR protein release, whereas PAR-2 AP shows no such effects. These results indicate that HAT enhances mucin gene expression through an AR-EGFR pathway, and PAR-2 is not sufficient for or does not directly cause HAT-induced mucin gene expression. Thus, HAT might be a possible therapeutic target to prevent excessive mucus production in patients with chronic airway diseases.
Sigma-Aldrich
Anti-EGFR Antibody, neutralizing, clone LA1, clone LA1, Upstate®, from mouse