MilliporeSigma
  • Human Neural Stem Cell Transplantation Rescues Functional Deficits in R6/2 and Q140 Huntington's Disease Mice.
Human Neural Stem Cell Transplantation Rescues Functional Deficits in R6/2 and Q140 Huntington's Disease Mice.
Stem cell reports (2017-12-14)
Jack C Reidling, Aroa Relaño-Ginés, Sandra M Holley, Joseph Ochaba, Cindy Moore, Brian Fury, Alice Lau, Andrew H Tran, Sylvia Yeung, Delaram Salamati, Chunni Zhu, Asa Hatami, Carlos Cepeda, Joshua A Barry, Talia Kamdjou, Alvin King, Dane Coleal-Bergum, Nicholas R Franich, Frank M LaFerla, Joan S Steffan, Mathew Blurton-Jones, Charles K Meshul, Gerhard Bauer, Michael S Levine, Marie-Francoise Chesselet, Leslie M Thompson
ABSTRACT
Huntington's disease (HD) is an inherited neurodegenerative disorder with no disease-modifying treatment. Expansion of the glutamine-encoding repeat in the Huntingtin (HTT) gene causes broad effects that are a challenge for single treatment strategies. Strategies based on human stem cells offer a promising option. We evaluated efficacy of transplanting a good manufacturing practice (GMP)-grade human embryonic stem cell-derived neural stem cell (hNSC) line into striatum of HD modeled mice. In HD fragment model R6/2 mice, transplants improve motor deficits, rescue synaptic alterations, and are contacted by nerve terminals from mouse cells. Furthermore, implanted hNSCs are electrophysiologically active. hNSCs also improved motor and late-stage cognitive impairment in a second HD model, Q140 knockin mice. Disease-modifying activity is suggested by the reduction of aberrant accumulation of mutant HTT protein and expression of brain-derived neurotrophic factor (BDNF) in both models. These findings hold promise for future development of stem cell-based therapies.
Sigma-Aldrich
Anti-Huntingtin Antibody, a.a. 1-82, ascites fluid, clone 2B4, Chemicon®