MilliporeSigma
  • Recombinant collagen scaffolds as substrates for human neural stem/progenitor cells.
Recombinant collagen scaffolds as substrates for human neural stem/progenitor cells.
Journal of biomedical materials research. Part A (2018-01-18)
Richard A Que, Janahan Arulmoli, Nancy A Da Silva, Lisa A Flanagan, Szu-Wen Wang
ABSTRACT
Adhesion to the microenvironment profoundly affects stem cell functions, including proliferation and differentiation, and understanding the interaction of stem cells with the microenvironment is important for controlling their behavior. In this study, we investigated the effects of the integrin binding epitopes GFOGER and IKVAV (natively present in collagen I and laminin, respectively) on human neural stem/progenitor cells (hNSPCs). To test the specificity of these epitopes, GFOGER or IKVAV were placed within the context of recombinant triple-helical collagen III engineered to be devoid of native integrin binding sites. HNSPCs adhered to collagen that presented GFOGER as the sole integrin-binding site, but not to IKVAV-containing collagen. For the GFOGER-containing collagens, antibodies against the β1 integrin subunit prevented cellular adhesion, antibodies against the α1 subunit reduced cell adhesion, and antibodies against α2 or α3 subunits had no significant effect. These results indicate that hNSPCs primarily interact with GFOGER through the α1β1 integrin heterodimer. These GFOGER-presenting collagen variants also supported differentiation of hNSPCs into neurons and astrocytes. Our findings show, for the first time, that hNSPCs can bind to the GFOGER sequence, and they provide motivation to develop hydrogels formed from recombinant collagen variants as a cell delivery scaffold. © 2018 Wiley Periodicals, Inc. J Biomed Mater Res Part A: 106A: 1363-1372, 2018.
Sigma-Aldrich
Anti-Integrin α3 Antibody, clone P1B5, azide free, clone P1B5, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin α1 Antibody, clone FB12, azide free, clone FB12, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin β1 Antibody, clone 6S6, Azide Free, clone 6S6, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin α2 Antibody, clone P1E6, azide free, clone P1E6, Chemicon®, from mouse