MilliporeSigma
  • Dielectrophoresis based discrimination of human embryonic stem cells from differentiating derivatives.
Dielectrophoresis based discrimination of human embryonic stem cells from differentiating derivatives.
Biomicrofluidics (2012-01-01)
Srinivas Velugotla, Steve Pells, Heidi K Mjoseng, Cairnan R E Duffy, Stewart Smith, Paul De Sousa, Ronald Pethig
ABSTRACT
Assessment of the dielectrophoresis (DEP) cross-over frequency (f xo), cell diameter, and derivative membrane capacitance (C m) values for a group of undifferentiated human embryonic stem cell (hESC) lines (H1, H9, RCM1, RH1), and for a transgenic subclone of H1 (T8) revealed that hESC lines could not be discriminated on their mean f xo and C m values, the latter of which ranged from 14 to 20 mF/m(2). Differentiation of H1 and H9 to a mesenchymal stem cell-like phenotype resulted in similar significant increases in mean C m values to 41-49 mF/m(2) in both lines (p < 0.0001). BMP4-induced differentiation of RCM1 to a trophoblast cell-like phenotype also resulted in a distinct and significant increase in mean C m value to 28 mF/m(2) (p < 0.0001). The progressive transition to a higher membrane capacitance was also evident after each passage of cell culture as H9 cells transitioned to a mesenchymal stem cell-like state induced by growth on a substrate of hyaluronan. These findings confirm the existence of distinctive parameters between undifferentiated and differentiating cells on which future application of dielectrophoresis in the context of hESC manufacturing can be based.
Sigma-Aldrich
Anti-STRO-1 Antibody, clone STRO-1, ascites fluid, clone STRO-1, Chemicon®